• USA Home
  • 276065 - (−)-N,N′-Dibenzyl-D-tartaric diamide

EMAIL THIS PAGE TO A FRIEND
276065 Sigma-Aldrich

(−)-N,N′-Dibenzyl-D-tartaric diamide

99%

Synonym: (−)-N,N′-Dibenzyl-D-tartramide

  • CAS Number 108321-43-3

  • Linear Formula [CH(OH)CONHCH2C6H5]2

  • Molecular Weight 328.36

  •  Beilstein/REAXYS Number 4551861

  •  MDL number MFCD00010269

  •  PubChem Substance ID 24856487

Purchase

Properties

Related Categories Amides, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
assay   99%
optical activity   [α]20/D −83°, c = 5.5 in pyridine
mp   198-200 °C (lit.)
SMILES string   O[C@@H]([C@H](O)C(=O)NCc1ccccc1)C(=O)NCc2ccccc2
InChI   1S/C18H20N2O4/c21-15(17(23)19-11-13-7-3-1-4-8-13)16(22)18(24)20-12-14-9-5-2-6-10-14/h1-10,15-16,21-22H,11-12H2,(H,19,23)(H,20,24)/t15-,16-/m0/s1
InChI key   BBYSAHVLSFBCMN-HOTGVXAUSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?