• USA Home
  • 276278 - Resorcinol diacetate

EMAIL THIS PAGE TO A FRIEND
276278 Sigma-Aldrich

Resorcinol diacetate

98%

Synonym: 1,3-Diacetoxybenzene

Purchase

Properties

assay   98%
refractive index   n20/D 1.503 (lit.)
bp   146 °C/12 mmHg(lit.)
density   1.178 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VH0580000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?