EMAIL THIS PAGE TO A FRIEND
278319 Sigma-Aldrich

Indium

wire, diam. 2.0 mm, 99.99% trace metals basis

Purchase

Properties

Related Categories Indium, Materials Science, Metal and Ceramic Science, Metals
vapor pressure   <0.01 mmHg ( 25 °C)
InChI Key   APFVFJFRJDLVQX-UHFFFAOYSA-N
assay   99.99% trace metals basis
form   wire
resistivity   8.37 μΩ-cm
diam.   2.0 mm
mp   156.6 °C (lit.)
density   7.3 g/mL at 25 °C(lit.)

Description

Quantity

5.6 g = 25 cm; 28 g = 125 cm

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NL1050000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?