• USA Home
  • 278807 - (S)-(+)-5-(1-Hydroxy-1-methylethyl)-2-methyl-2-cyclohexen-1-one

EMAIL THIS PAGE TO A FRIEND
278807 Sigma-Aldrich

(S)-(+)-5-(1-Hydroxy-1-methylethyl)-2-methyl-2-cyclohexen-1-one

98%

Synonym: (S)-(+)-Carvone hydrate

  • CAS Number 60593-11-5

  • Empirical Formula (Hill Notation) C10H16O2

  • Molecular Weight 168.23

  •  PubChem Substance ID 24856833

Purchase

Properties

assay   98%
optical activity   [α]19/D +41°, c = 10 in ethanol
refractive index   n20/D 1.507 (lit.)
bp   157-158 °C/14 mmHg (lit.)
mp   40-42 °C (lit.)
density   1.043 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?