• USA Home
  • 280089 - 1-Bromo-2,3-dichlorobenzene

EMAIL THIS PAGE TO A FRIEND
280089 Sigma-Aldrich

1-Bromo-2,3-dichlorobenzene

98%

Purchase

Properties

assay   98%
bp   243 °C/765 mmHg (lit.)
mp   58-61 °C (lit.)
SMILES string   Clc1cccc(Br)c1Cl
InChI   1S/C6H3BrCl2/c7-4-2-1-3-5(8)6(4)9/h1-3H
InChI key   HVKCZUVMQPUWSX-UHFFFAOYSA-N

Description

Packaging

100 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?