• USA Home
  • 280739 - Tetraisopropyl orthocarbonate

EMAIL THIS PAGE TO A FRIEND
280739

Tetraisopropyl orthocarbonate

97%

  • CAS Number 36597-49-6

  • Empirical Formula (Hill Notation) C13H28O4

  • Molecular Weight 248.36

  •  MDL number MFCD00008878

Purchase

Properties

assay   97%
SMILES string   CC(C)OC(OC(C)C)(OC(C)C)OC(C)C
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?