EMAIL THIS PAGE TO A FRIEND
282057 Aldrich

Sodium

99%, lumps, contains kerosene

Purchase

Properties

Related Categories Alkali Metals, Chemical Synthesis, Materials Science, Metal and Ceramic Science, Metals,
vapor pressure   1 mmHg ( 440 °C)
  1 mmHg ( 440 °C)
InChI Key   KEAYESYHFKHZAL-UHFFFAOYSA-N
assay   99%
form   lumps
contains   kerosene
resistivity   4.69 μΩ-cm, 20°C
bp   883 °C(lit.)
mp   97.8 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1428 4.3 / PGI
WGK Germany 
3
RTECS 
VY0686000
Flash Point(F) 
179.6 °F
Flash Point(C) 
82 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?