• USA Home
  • 282324 - 1,3,5-Trichloro-2,4,6-trifluorobenzene

EMAIL THIS PAGE TO A FRIEND
282324 Sigma-Aldrich

1,3,5-Trichloro-2,4,6-trifluorobenzene

98%

Purchase

Properties

assay   98%
bp   79.5 °C/12 mmHg(lit.)
mp   62-64 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DC2450000
Flash Point(F) 
215.6 °F
Flash Point(C) 
102 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?