• USA Home
  • 282340 - Barium pentafluorobenzenesulfonate

EMAIL THIS PAGE TO A FRIEND
282340 Sigma-Aldrich

Barium pentafluorobenzenesulfonate

Synonym: Pentafluorobenzenesulfonic acid barium salt

Purchase
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?