• USA Home
  • 282545 - 7-Amino-4-hydroxy-2-naphthalenesulfonic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
282545 Sigma-Aldrich

7-Amino-4-hydroxy-2-naphthalenesulfonic acid monohydrate

97%

Purchase

Properties

assay   97%
SMILES string   O.Nc1ccc2c(O)cc(cc2c1)S(O)(=O)=O
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2585PSN2 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

08800 7-Amino-4-hydroxy-2-naphthalenesulfonic acid, technical, ≥90% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?