EMAIL THIS PAGE TO A FRIEND
282596 Sigma-Aldrich

1-Hexacosanol

synthetic, 98%

Synonym: Ceryl alcohol, Hexacosyl alcohol

  • CAS Number 506-52-5

  • Linear Formula CH3(CH2)25OH

  • Molecular Weight 382.71

  •  Beilstein/REAXYS Number 1783162

  •  MDL number MFCD00002940

Purchase

Properties

assay   98%
SMILES string   CCCCCCCCCCCCCCCCCCCCCCCCCCO
InChI   1S/C26H54O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27/h27H,2-26H2,1H3
InChI key   IRHTZOCLLONTOC-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?