• USA Home
  • 282685 - 1-Acetyl-2-methyl-1-cyclopentene

EMAIL THIS PAGE TO A FRIEND
282685 Sigma-Aldrich

1-Acetyl-2-methyl-1-cyclopentene

96%

  • CAS Number 3168-90-9

  • Empirical Formula (Hill Notation) C8H12O

  • Molecular Weight 124.18

  •  MDL number MFCD00010610

Purchase

Properties

assay   96%
SMILES string   CC(=O)C1=C(C)CCC1
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
152.6 °F
Flash Point(C) 
67 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?