• USA Home
  • 283347 - 1,2,4,5-Tetramethyl-1,4-cyclohexadiene

EMAIL THIS PAGE TO A FRIEND
283347 Sigma-Aldrich

1,2,4,5-Tetramethyl-1,4-cyclohexadiene

98%

  • CAS Number 26976-92-1

  • Empirical Formula (Hill Notation) C10H16

  • Molecular Weight 136.23

  •  MDL number MFCD00010373

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?