• USA Home
  • 284114 - Promethazine hydrochloride

EMAIL THIS PAGE TO A FRIEND
284114 Aldrich

Promethazine hydrochloride

98%

Synonym: 10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride

  • CAS Number 58-33-3

  • Empirical Formula (Hill Notation) C17H20N2S · HCl

  • Molecular Weight 320.88

  •  Beilstein/REAXYS Number 4166397

  •  EC Number 200-375-2

  •  MDL number MFCD00012652

Purchase

Properties

InChI Key   XXPDBLUZJRXNNZ-UHFFFAOYSA-N
assay   98%
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
SO8225000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?