EMAIL THIS PAGE TO A FRIEND
284157 Sigma-Aldrich

Oleic anhydride

98%

Synonym: cis-9-Octadecenoic anhydride

  • CAS Number 24909-72-6

  • Linear Formula [CH3(CH2)7CH=CH(CH2)7CO]2O

  • Molecular Weight 546.91

  •  Beilstein/REAXYS Number 1730211

  •  EC Number 246-522-4

  •  MDL number MFCD00010459

Purchase

Properties

InChI Key   OCNZHGHKKQOQCZ-CLFAGFIQSA-N
assay   98%
bp   200-215 °C/11 mmHg (lit.)
mp   22-24 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

75095 Oleic anhydride, ≥95.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?