EMAIL THIS PAGE TO A FRIEND
284289 Sigma-Aldrich

Hexacosanoic acid

98%

Synonym: Cerotic acid, Cerotinic acid

  • CAS Number 506-46-7

  • Linear Formula CH3(CH2)24COOH

  • Molecular Weight 396.69

  •  Beilstein/REAXYS Number 1799681

  •  MDL number MFCD00002811

Purchase

Properties

assay   98%
SMILES string   CCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
InChI   1S/C26H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28/h2-25H2,1H3,(H,27,28)
InChI key   XMHIUKTWLZUKEX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?