• USA Home
  • 285129 - 4-Androsten-19-al-3,17-dione

EMAIL THIS PAGE TO A FRIEND
285129 Sigma-Aldrich

4-Androsten-19-al-3,17-dione

97%

Synonym: 19-Aldoandrostenedione

  • CAS Number 968-49-0

  • Empirical Formula (Hill Notation) C19H24O3

  • Molecular Weight 300.39

  •  MDL number MFCD00010471

Purchase

Properties

assay   97%
SMILES string   C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C=O)[C@@H]1CCC2=O

Description

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 285129.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?