• USA Home
  • 285307 - Cephaloridine hydrate

EMAIL THIS PAGE TO A FRIEND
285307 Sigma-Aldrich

Cephaloridine hydrate

98%

  • CAS Number 102039-86-1

  • Empirical Formula (Hill Notation) C19H17N3O4S2 · xH2O

  • Molecular Weight 415.49 (anhydrous basis)

  •  EC Number 200-052-6

  •  MDL number MFCD00150011

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
UU3675000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?