• USA Home
  • 285560 - Diethylenetriaminepentaacetic acid

EMAIL THIS PAGE TO A FRIEND
285560 Sigma-Aldrich

Diethylenetriaminepentaacetic acid

98%

Synonym: (Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid

  • CAS Number 67-43-6

  • Linear Formula [(HOOCCH2)2NCH2CH2]2NCH2COOH

  • Molecular Weight 393.35

  •  Beilstein/REAXYS Number 1810219

  •  EC Number 200-652-8

  •  MDL number MFCD00004289

Purchase

Properties

InChI Key   QPCDCPDFJACHGM-UHFFFAOYSA-N
assay   98%
mp   219-220 °C (lit.)

Description

Application

Heavy metal chelating agent

Safety & Documentation

Safety Information

RIDADR 
UN 3077 9/PG 3
WGK Germany 
2
RTECS 
MB8205000
Flash Point(F) 
392 °F
Flash Point(C) 
200 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D6518 Diethylenetriaminepentaacetic acid, ≥99% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?