EMAIL THIS PAGE TO A FRIEND
285676 Sigma-Aldrich

Dipyridamole

98%

Purchase

Properties

InChI Key   IZEKFCXSFNUWAM-UHFFFAOYSA-N
assay   98%
mp   165-166 °C (lit.)
Gene Information   human ... CYP1A2(1544), PDE10A(10846), SLC29A1(2030)
mouse ... Slc29a1(63959)

Description

Biochem/physiol Actions

Selective inhibitor of phosphodiesterase V (PDE 5); potent coronary vasodilator drug; adenosine transport inhibitor; inhibitor of platelet aggregation.

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
KK7450000

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D9766 Dipyridamole, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?