• USA Home
  • 285935 - Gallamine triethiodide

EMAIL THIS PAGE TO A FRIEND
285935 Aldrich

Gallamine triethiodide

98%

Purchase

Properties

InChI Key   REEUVFCVXKWOFE-UHFFFAOYSA-K
assay   98%
mp   235 °C (dec.) (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
BS1100000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

G8134 Gallamine triethiodide, ≥98% (TLC), powder, muscarinic receptor antagonist

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?