• USA Home
  • 286206 - 11β-Hydroxyprogesterone

EMAIL THIS PAGE TO A FRIEND
286206 Aldrich

11β-Hydroxyprogesterone

98%

Synonym: 11β-Hydroxy-4-pregnene-3,20-dione, 11β-Hydroxypregn-4-ene-3,20-dione, 4-Pregnen-11β-ol-3,20-dione

  • CAS Number 600-57-7

  • Empirical Formula (Hill Notation) C21H30O3

  • Molecular Weight 330.46

  •  MDL number MFCD00064137

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?