• USA Home
  • 286648 - Medroxyprogesterone 17-acetate

EMAIL THIS PAGE TO A FRIEND
286648 Aldrich

Medroxyprogesterone 17-acetate

97%

Synonym: 17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate

  • CAS Number 71-58-9

  • Empirical Formula (Hill Notation) C24H34O4

  • Molecular Weight 386.52

  •  Beilstein/REAXYS Number 2066112

  •  EC Number 200-757-9

  •  MDL number MFCD00010483

Purchase

Properties

InChI Key   PSGAAPLEWMOORI-PEINSRQWSA-N
assay   97%
optical activity   [α]25/D +58°, c = 1 in chloroform
mp   206-207 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
TU5010000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M1629 Medroxyprogesterone 17-acetate, ≥97% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?