• USA Home
  • 286745 - Nalidixic acid sodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
286745 Sigma-Aldrich

Nalidixic acid sodium salt hydrate

97%

Synonym: 1,4-Dihydro-1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt

  • CAS Number 15769-77-4

  • Empirical Formula (Hill Notation) C12H11N2NaO3 · xH2O

  • Molecular Weight 254.22 (anhydrous basis)

  •  EC Number 222-159-7

  •  MDL number MFCD00150870

  •  PubChem Substance ID 24857249

Purchase

Properties

assay   97%
impurities   ≤1% ethyl alcohol

Description

Other Notes

Contains acetone

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QN2885500
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?