EMAIL THIS PAGE TO A FRIEND
287199 Sigma-Aldrich

Sulfadiazine

99%

Synonym: 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide, N1-(Pyrimidin-2-yl)sulfanilamide

  • CAS Number 68-35-9

  • Empirical Formula (Hill Notation) C10H10N4O2S

  • Molecular Weight 250.28

  •  Beilstein/REAXYS Number 6733588

  •  EC Number 200-685-8

  •  MDL number MFCD00006065

Purchase

Properties

InChI Key   SEEPANYCNGTZFQ-UHFFFAOYSA-N
assay   99%
mp   253 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
WP1925000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S8626 Sulfadiazine, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?