• USA Home
  • 288195 - Phenylgermanium trichloride

EMAIL THIS PAGE TO A FRIEND
288195 Sigma-Aldrich

Phenylgermanium trichloride

98%

Purchase

Properties

Related Categories Chemical Synthesis, Organogermanium, Organometallic Reagents More...
assay   98%
refractive index   n20/D 1.554 (lit.)
bp   105-106 °C/12 mmHg (lit.)
density   1.584 g/mL at 25 °C (lit.)
SMILES string   Cl[Ge](Cl)(Cl)c1ccccc1
InChI   1S/C6H5Cl3Ge/c7-10(8,9)6-4-2-1-3-5-6/h1-5H
InChI key   CIWQSBMDFPABPQ-UHFFFAOYSA-N

Description

Packaging

1, 5 g in ampule

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?