EMAIL THIS PAGE TO A FRIEND
288675 Sigma-Aldrich

Sodium selenate

98%

Purchase

Properties

assay   98%
SMILES string   [Na+].[Na+].[O-][Se]([O-])(=O)=O
InChI   1S/2Na.H2O4Se/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key   MHQOTKLEMKRJIR-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2630PSN1 6.1 / PGI
WGK Germany 
2
RTECS 
VS6650000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?