• USA Home
  • 28964 - (S)-1-Benzyl-3-hydroxypiperidine

EMAIL THIS PAGE TO A FRIEND
28964 Sigma-Aldrich

(S)-1-Benzyl-3-hydroxypiperidine

≥97.0% (GC)

Synonym: (S)-1-Benzyl-3-piperidinol

Purchase

Properties

assay   ≥97.0% (GC)
optical activity   [α]20/D 13.5±1.5°, c = 2.1 in methanol
refractive index   n20/D 1.545±0.002
storage temp.   2-8°C
SMILES string   O[C@H]1CCCN(C1)Cc2ccccc2

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
2
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?