• USA Home
  • 289701 - N,N-Dimethyldodecylamine N-oxide

EMAIL THIS PAGE TO A FRIEND
289701 Aldrich

N,N-Dimethyldodecylamine N-oxide

98%

Synonym: DDAO, LDAO, Lauryldimethylamine N-oxide

  • CAS Number 1643-20-5

  • Linear Formula CH3(CH2)11N(O)(CH3)2

  • Molecular Weight 229.40

  •  Beilstein Registry Number 1769927

  •  EC Number 216-700-6

  •  MDL number MFCD00002049

Purchase

Properties

InChI Key   SYELZBGXAIXKHU-UHFFFAOYSA-N
assay   98%
mp   132-133 °C (lit.)

Description

Application

DDAO is a non-denaturing zwitterionic surfactant with a critical micelle concentration (CMC) or approximately 1 mM. It is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3259PSN1 8 / PGII
WGK Germany 
2
RTECS 
JR6650000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?