• USA Home
  • 289949 - Potassium L-tartrate

EMAIL THIS PAGE TO A FRIEND
289949 Sigma-Aldrich

Potassium L-tartrate

97%

Synonym: L-Tartaric acid dipotassium salt

Purchase

Properties

assay   97%
optical activity   [α]20/D +26°, c = 1 in H2O
SMILES string   [K+].[K+].[H]O[H].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
InChI   1S/C4H6O6.2K.H2O/c5-1(3(7)8)2(6)4(9)10;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;1H2/q;2*+1;/p-2/t1-,2-;;;/m1.../s1
InChI key   BUCMCXRUIRJPKS-QXMMYYGRSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?