• USA Home
  • 290041 - (1S,2S)-(+)-N-Methylpseudoephedrine

EMAIL THIS PAGE TO A FRIEND
290041 Sigma-Aldrich

(1S,2S)-(+)-N-Methylpseudoephedrine

99%

Synonym: (+)-N-Methylpseudoephedrine, (1S,2S)-2-Dimethylamino-1-phenylpropanol

Purchase

Properties

assay   99%
optical activity   [α]21/D +48°, c = 5 in methanol
bp   145 °C/24 mmHg (lit.)
mp   29-31 °C (lit.)
storage temp.   2-8°C
SMILES string   C[C@@H]([C@@H](O)c1ccccc1)N(C)C
InChI   1S/C11H17NO/c1-9(12(2)3)11(13)10-7-5-4-6-8-10/h4-9,11,13H,1-3H3/t9-,11+/m0/s1
InChI key   FMCGSUUBYTWNDP-GXSJLCMTSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?