• USA Home
  • 291285 - α-Hexylcinnamaldehyde

EMAIL THIS PAGE TO A FRIEND
291285 Sigma-Aldrich

α-Hexylcinnamaldehyde

technical grade, 85%

Synonym: Hexyl cinnamal, alpha-Hexylcinnamaldehyde

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C13-C60, Carbonyl Compounds, Chemical Synthesis,
grade   technical grade
InChI Key   GUUHFMWKWLOQMM-NTCAYCPXSA-N
assay   85%
contains   0.3 ppm butylated hydroxyanisole as stabilizer
refractive index   n20/D 1.55 (lit.)
bp   174-176 °C/15 mmHg (lit.)
solubility   water: 0.005 g/L at 25 °C
density   0.95 g/mL at 25 °C (lit.)

Description

General description

Solubilization of α-hexylcinnamaldehyde by the ionic surfactants, sodium dodecyl sulfate and dodecyltrimethylammonium chloride and a non-ionic surfactant such as dodecyl polyoxyethylene ethers has been investigated.

Application

α-Hexylcinnamaldehyde is characterized by a typical floral scent, which makes them suitable to be used as fragrances in personal care (perfumes, creams, shampoos, etc.) and household products. It has been also used as flavouring additive in food and pharmaceutical industry.

Packaging

250 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Articles

GC Analysis of Fragrance Compounds in Shampoo Base on a 100% Methyl Column after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Fragrance Compounds in Shampoo Base on a 100% Methyl Column after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Cosmetics, Gas chromatography, Mass selective detector, Purification, Solid phase microextractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?