• USA Home
  • 292117 - 2-Methoxy-1-pyrrolidinecarboxaldehyde,mixture of cis and trans

EMAIL THIS PAGE TO A FRIEND
292117

2-Methoxy-1-pyrrolidinecarboxaldehyde,mixture of cis and trans

97%

  • CAS Number 61020-06-2

  • Empirical Formula (Hill Notation) C6H11NO2

  • Molecular Weight 129.16

  •  MDL number MFCD00010850

Purchase

Properties

assay   97%
SMILES string   COC1CCCN1C=O
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?