• USA Home
  • 292400 - 1-Deoxy-D-gluco-hex-1-enopyranose tetrabenzoate

EMAIL THIS PAGE TO A FRIEND
292400 Aldrich

1-Deoxy-D-gluco-hex-1-enopyranose tetrabenzoate

  • CAS Number 14125-75-8

  • Empirical Formula (Hill Notation) C34H26O9

  • Molecular Weight 578.56

  •  PubChem Substance ID 24857583

Purchase

Properties

optical activity   [α]18/D −88°, c = 1 in chloroform
mp   122-124 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?