EMAIL THIS PAGE TO A FRIEND
293229 Supelco

1-Chlorobutane

HPLC grade, ≥99.5%

Synonym: Butyl chloride

Purchase

Properties

grade   HPLC grade
vapor density   3.2 (vs air)
vapor pressure   80.1 mmHg ( 78.4 °C)
InChI Key   VFWCMGCRMGJXDK-UHFFFAOYSA-N
assay   ≥99.5%
autoignition temp.   860 °F
purified by   glass distillation
expl. lim.   10.1 %
impurities   <0.020% water
evapn. residue   <0.0003%
refractive index   n20/D 1.402 (lit.)
bp   77-78 °C (lit.)
  77-78 °C
mp   −123 °C (lit.)
density   0.886 g/mL at 25 °C (lit.)
λ   H2O reference
UV absorption   λ: 220 nm Amax: 1.00
  λ: 225 nm Amax: 0.35
  λ: 250 nm Amax: 0.05
  λ: 300-400 nm Amax: 0.01
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1127 - class 3 - PG 2 - Chlorobutanes
WGK Germany 
2
RTECS 
EJ6300000
Flash Point(F) 
10.4 °F
Flash Point(C) 
-12 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?