• USA Home
  • 293563 - Dehydroabietylamine acetate

EMAIL THIS PAGE TO A FRIEND
293563 Sigma-Aldrich

Dehydroabietylamine acetate

technical grade, 85%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Complex Molecules
grade   technical grade
assay   85%
optical activity   [α]20/D +50°, c = 2.5 in pyridine
mp   139-141 °C (lit.)
SMILES string   CC(O)=O.[H][C@@]12CCc3cc(ccc3[C@@]1(C)CCC[C@@]2(C)CN)C(C)C
InChI   1S/C20H31N.C2H4O2/c1-14(2)15-6-8-17-16(12-15)7-9-18-19(3,13-21)10-5-11-20(17,18)4;1-2(3)4/h6,8,12,14,18H,5,7,9-11,13,21H2,1-4H3;1H3,(H,3,4)/t18-,19-,20+;/m0./s1
InChI key   BFAQRUGPWJVQDA-WFBUOHSLSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
SF7750000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?