• USA Home
  • 293695 - (4-Nitrobenzyl)trimethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
293695 Sigma-Aldrich

(4-Nitrobenzyl)trimethylammonium chloride

98%

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   SBPSWIPEYIVRNF-UHFFFAOYSA-M
assay   98%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?