EMAIL THIS PAGE TO A FRIEND
293903 Sigma-Aldrich

Diisooctyl azelate

technical grade

Purchase

Properties

grade   technical grade
description   mixture of C8 isomers
refractive index   n20/D 1.451 (lit.)
bp   208-210 °C/2 mmHg (lit.)
density   0.905 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
-
RTECS 
CM2000000
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?