• USA Home
  • 293997 - 5-DOXYL-decane, free radical

EMAIL THIS PAGE TO A FRIEND
293997 Sigma-Aldrich

5-DOXYL-decane, free radical

Synonym: 2-Butyl-4,4-dimethyl-2-pentyl-3-oxazolidinyloxy, free radical

  • CAS Number 35203-80-6

  • Empirical Formula (Hill Notation) C14H29NO2

  • Molecular Weight 243.39

  •  MDL number MFCD00011559

Purchase

Properties

SMILES string   CCCCCC1(CCCC)OCC(C)(C)N1[O]
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?