• USA Home
  • 294608 - 3-Amino-2-butenethioamide

EMAIL THIS PAGE TO A FRIEND
294608 Sigma-Aldrich

3-Amino-2-butenethioamide

98%

Purchase

Properties

assay   98%
mp   135-138 °C (lit.)
SMILES string   C\C(N)=C/C(N)=S
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?