• USA Home
  • 29529 - (S)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis{bis[3,5-diisopropyl-4-(dimethylamino)phenyl]phosphine}

EMAIL THIS PAGE TO A FRIEND
29529 Sigma-Aldrich

(S)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis{bis[3,5-diisopropyl-4-(dimethylamino)phenyl]phosphine}

≥97%, optical purity ee: ≥99%

Synonym: (S)-2,2′-Bis[bis(3,5-diisopropyl-4-dimethylaminophenyl)phosphino]-6,6′-dimethoxy-1,1′-biphenyl, (S)-3,5-iPr-4-NMe2-MeOBIPHEP, SL-A107-2

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, MeOBIPHEP Ligands, Privileged Ligands and Complexes,
InChI Key   SBQGDYGNDIQAIT-UHFFFAOYSA-N
assay   ≥97%
optical purity   ee: ≥99%

Description

General description

sold in collaboration with Solvias AG

Application

Atropisomeric MeOBIPHEP ligands

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Solvias® Ligand Portfolio for Enantioselective Hydrogenation

Today's Solvias Ligand Portfolio has its roots in the development of the Josiphos ligands and the successful implementation in the (S)-metolachlor process—the largest industrial enantioselective manu...
William Sommer and Daniel Weibel
Aldrich ChemFiles 2008, 8.2, 14.
Keywords: Alkylations, Aminations, Asymmetric catalysis, Asymmetric synthesis, Building blocks, Catalysis, Coupling reactions, Crystallization, Grignard Reaction, Heck Reaction, Herbicides, Hydroformylations, Hydrogenations, Infrared spectroscopy, Kumada Coupling, Ligands, Michael Addition, Reductions, Supercritical fluid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?