• USA Home
  • 295779 - 1,8-Diaminonaphthalene

EMAIL THIS PAGE TO A FRIEND
295779

1,8-Diaminonaphthalene

technical grade, 90%

Synonym: 1,8-Naphthalenediamine

  • CAS Number 479-27-6

  • Linear Formula C10H6(NH2)2

  • Molecular Weight 158.20

  •  Beilstein/REAXYS Number 742841

  •  MDL number MFCD00004033

Purchase

Properties

grade   technical grade
assay   90%
bp   205 °C/12 mmHg (lit.)
mp   60-65 °C (lit.)
SMILES string   Nc1cccc2cccc(N)c12
InChI   1S/C10H10N2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H,11-12H2
InChI key   YFOOEYJGMMJJLS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?