• USA Home
  • 296228 - N-Acetyl-1,2-diphenylhydrazine

EMAIL THIS PAGE TO A FRIEND
296228

N-Acetyl-1,2-diphenylhydrazine

99%

  • CAS Number 22293-38-5

  • Empirical Formula (Hill Notation) C14H14N2O

  • Molecular Weight 226.27

  •  MDL number MFCD00010714

Purchase

Properties

assay   99%
SMILES string   CC(=O)N(Nc1ccccc1)c2ccccc2
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?