• USA Home
  • 296252 - 4-Amino-3-chloro-2,5,6-trifluoropyridine

EMAIL THIS PAGE TO A FRIEND
296252 Sigma-Aldrich

4-Amino-3-chloro-2,5,6-trifluoropyridine

99%

  • CAS Number 2693-57-4

  • Empirical Formula (Hill Notation) C5H2ClF3N2

  • Molecular Weight 182.53

  •  MDL number MFCD00010856

  •  PubChem Substance ID 24857846

Purchase

Properties

Related Categories Building Blocks, C5, Chemical Synthesis, Fluorinated Building Blocks, Halogenated Heterocycles,
assay   99%
mp   120-121 °C (lit.)
SMILES string   Nc1c(F)c(F)nc(F)c1Cl
InChI   1S/C5H2ClF3N2/c6-1-3(10)2(7)5(9)11-4(1)8/h(H2,10,11)
InChI key   IJGMBUFHHBUVNG-UHFFFAOYSA-N

Description

Application

4-Amino-3-chloro-2,5,6-trifluoropyridine may be used in chemical synthesis.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?