• USA Home
  • 296414 - Phenyl β-D-galactopyranoside

EMAIL THIS PAGE TO A FRIEND
296414 Aldrich

Phenyl β-D-galactopyranoside

98%

Synonym: P-Gal, Phenyl β-D-galactoside

  • CAS Number 2818-58-8

  • Empirical Formula (Hill Notation) C12H16O6

  • Molecular Weight 256.25

  •  Beilstein/REAXYS Number 87518

  •  EC Number 220-573-2

  •  MDL number MFCD00063258

Purchase

Properties

assay   98%
optical activity   [α]25/D −40°, c = 2.3 in H2O
mp   153-155 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P6501 Phenyl-β-D-galactopyranoside, ≥98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?