• USA Home
  • 296430 - (−)-Dimethyl 2,3-O-benzylidene-L-tartrate

EMAIL THIS PAGE TO A FRIEND
296430 Sigma-Aldrich

(−)-Dimethyl 2,3-O-benzylidene-L-tartrate

99%

Synonym: (4R,5R)-2-Phenyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, Dimethyl (4R,5R)-2-phenyl-1,3-dioxolane-4,5-dicarboxylate

  • CAS Number 38270-72-3

  • Empirical Formula (Hill Notation) C13H14O6

  • Molecular Weight 266.25

  •  Beilstein/REAXYS Number 33816

  •  MDL number MFCD00064474

Purchase

Properties

assay   99%
optical activity   [α]20/D −41.0°, c = 2 in ethanol
mp   74-76 °C (lit.)
SMILES string   COC(=O)[C@@H]1OC(O[C@H]1C(=O)OC)c2ccccc2
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?