• USA Home
  • 296597 - Europium tris[3-(trifluoromethylhydroxymethylene)-(−)-camphorate]

EMAIL THIS PAGE TO A FRIEND
296597 Sigma-Aldrich

Europium tris[3-(trifluoromethylhydroxymethylene)-(−)-camphorate]

  • Empirical Formula (Hill Notation) C36H42EuF9O6

  • Molecular Weight 893.66

  •  EC Number 252-232-9

  •  MDL number MFCD00074803

  •  PubChem Substance ID 24857448

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Resolution Reagents, Chiral Shift Reagents, NMR - Buffers and Reagents,
InChI Key   BWIHMSGCCRUUCZ-NCJHBDPTSA-K
optical activity   [α]18/D −156°, c = 1 in chloroform
mp   195-198 °C (lit.)

Description

Application

Optically active NMR shift reagent

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?