• USA Home
  • 296694 - (S)-(+)-3-Methyl-2-butanol

EMAIL THIS PAGE TO A FRIEND
296694 Aldrich

(S)-(+)-3-Methyl-2-butanol

98%

  • CAS Number 1517-66-4

  • Empirical Formula (Hill Notation) C5H12O

  • Molecular Weight 88.15

  •  MDL number MFCD00064271

Purchase

Properties

assay   98%
optical purity   ee: 96% (GLC)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1105 - class 3 - PG 3 - Pentanols
WGK Germany 
3
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?