• USA Home
  • 297976 - Tributylborane solution

EMAIL THIS PAGE TO A FRIEND
297976 Sigma-Aldrich

Tributylborane solution

1.0 M in diethyl ether

Synonym: TBB, Tributylboron

  • CAS Number 122-56-5

  • Linear Formula [CH3(CH2)3]3B

  • Molecular Weight 182.15

  •  MDL number MFCD00009423

  •  PubChem Substance ID 24857950

  •  NACRES NA.22

Purchase

Properties

Related Categories Boranes, Chemical Synthesis, Reduction, Synthetic Reagents
Quality Level   100
concentration   1.0 M in diethyl ether
density   0.719 g/mL at 25 °C
SMILES string   CCCCB(CCCC)CCCC
InChI   1S/C12H27B/c1-4-7-10-13(11-8-5-2)12-9-6-3/h4-12H2,1-3H3
InChI key   CMHHITPYCHHOGT-UHFFFAOYSA-N

Description

Application

Reactant for:
• Addition reactions for synthesis of phenyltrialkylborate salts
• Boron-based multicomponent reactions for generation of substituted aziridines, oxazolidines, and pyrrolidines
• Suzuki-Miyaura cross coupling of halopurines with trialkylboranes for the synthesis of adenosine A2A receptor antagonists
• Multicomponent C-alkylation reactions with aromatic aldimines
• One-pot alkylation-boration and electrophilic substitution

Packaging

100 mL in Sure/Seal™

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Central nervous system
Supplemental Hazard Statements 
EUH019
RIDADR 
UN 1993A 3 / PGI
WGK Germany 
WGK 1
Flash Point(F) 
-40.0 °F - closed cup
Flash Point(C) 
-40 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?