• USA Home
  • 298565 - 7-Acetoxy-4-(bromomethyl)coumarin

EMAIL THIS PAGE TO A FRIEND
298565 Aldrich

7-Acetoxy-4-(bromomethyl)coumarin

97%

Synonym: 4-(Bromomethyl)umbelliferyl acetate, 7-Acetoxy-4-(bromomethyl)coumarin

  • CAS Number 2747-04-8

  • Empirical Formula (Hill Notation) C12H9BrO4

  • Molecular Weight 297.10

  •  Beilstein/REAXYS Number 220616

  •  MDL number MFCD00010707

Purchase

Properties

assay   97%
mp   180-182 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?